1-Bromo-3-n-octylbenzene ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
B4817-1G
B4817-5G
TCIAB4817-1GEA
765
SEK
TCIAB4817-1G
TCIAB4817-5G
1-Bromo-3-n-octylbenzene ≥97.0%
1-Bromo-3-n-octylbenzene
Formel:
C₁₄H₂₁Br MW: 269,23 g/mol |
CAS nummer:
663949-34-6 |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...