1-Bromocarbazole ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1-Bromo-9H-carbazole

B4816-200MG B4816-1G
TCIAB4816-200MGEA 310 SEK
TCIAB4816-200MG TCIAB4816-1G
1-Bromocarbazole ≥98.0%
1-Bromocarbazole
Formel: C₁₂H₈BrN
MW: 246,11 g/mol
MDL Number: MFCD18450164
CAS nummer: 16807-11-7

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 122.0 to 126.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR