3,9'-Bicarbazole ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
9H-3,9'-Bicarbazole
B4811-1G
B4811-5G
TCIAB4811-1GEA
935
SEK
TCIAB4811-1G
TCIAB4811-5G
3,9'-Bicarbazole ≥98.0% (by HPLC, total nitrogen)
3,9'-Bicarbazole
Formel:
C₂₄H₁₆N₂ MW: 332,41 g/mol |
MDL Number:
MFCD27644690 CAS nummer: 18628-07-4 |
Specifikation testresultat
Appearance | White to Gray to Red powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 214.0 to 218.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...