4-Bromo-4'-methylbiphenyl ≥98.0%
Leverantör: TCI
B4804-1G
B4804-5G
TCIAB4804-1GEA
1450
SEK
TCIAB4804-1G
TCIAB4804-5G
4-Bromo-4'-methylbiphenyl ≥98.0%
4-Bromo-4'-methylbiphenyl
Formel:
C₁₃H₁₁Br Kokpunkt: 315°C |
MDL Number:
MFCD00466236 CAS nummer: 50670-49-0 |
|
Specifikation testresultat
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 131.0 to 135.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...