N,O-Di-boc-hydroxylamine 98,0 GC_ANALYSMETOD

Leverantör: TCI
Varning

Synonyms: N,O-Di-Boc-hydroxylamine, tert-Butyl N-(tert-Butoxycarbonyloxy)carbamate, N,O-Bis(tert-butoxycarbonyl)hydroxylamine, tert-Butyl (tert-butoxycarbonyl)oxycarbamate, N-(tert-Butoxycarbonyloxy)carbamic acid tert-Butyl ester

B4780-1G B4780-5G
TCIAB4780-1GEA 285 SEK
TCIAB4780-1G TCIAB4780-5G
N,O-Di-boc-hydroxylamine 98,0 GC_ANALYSMETOD
N,O-Di-boc-hydroxylamine
Formel: C₁₀H₁₉NO₅
MW: 233.264 g/mol
Storage Temperature: Frys
MDL Number: MFCD00034797
CAS nummer: 85006-25-3
EINECS: 285-055-0

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 67.0 to 71.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR