4-Bromo-2-fluoropyridine ≥98.0%

Leverantör: TCI
Varning

B4778-1G B4778-5G
TCIAB4778-1GEA 695 SEK
TCIAB4778-1G TCIAB4778-5G
4-Bromo-2-fluoropyridine ≥98.0%
4-Bromo-2-fluoropyridine
Formel: C₅H₃BrFN
MW: 175.99 g/mol
Kokpunkt: 65°C
Flampunkt: 71°C
MDL Number: MFCD04112504
CAS nummer: 128071-98-7

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR