3-Benzyloxy-5-bromopyridine ≥98.0% (by GC, titration analysis)

Leverantör: TCI

B4775-1G B4775-5G
TCIAB4775-1GEA 415 SEK
TCIAB4775-1G TCIAB4775-5G
3-Benzyloxy-5-bromopyridine ≥98.0% (by GC, titration analysis)
3-Benzyloxy-5-bromopyridine
Formel: C₁₂H₁₀BrNO
MDL Number: MFCD07375004
CAS nummer: 130722-95-1

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 69.0 to 73.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR