3,5-Bis(trifluoromethyl)-1H-pyrazole ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 3,5-Bis(trifluoromethyl)pyrazole
B4762-1G B4762-5G
TCIAB4762-1GEA 575 SEK
TCIAB4762-1G TCIAB4762-5G
3,5-Bis(trifluoromethyl)-1H-pyrazole ≥98.0%
3,5-Bis(trifluoromethyl)-1H-pyrazole
Formel: C₅H₂F₆N₂
MW: 204,07 g/mol
Kokpunkt: 147°C
MDL Number: MFCD00153672
CAS nummer: 14704-41-7
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 83.0 to 87.0 deg-C