3'-Brom-4'-methyloacetophenone ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
3'-Bromo-4'-methylacetophenone, 1-(3-Bromo-4-methylphenyl)ethanone
B4758-1G
B4758-5G
TCIAB4758-1GEA
310
SEK
TCIAB4758-1G
TCIAB4758-5G
3'-Brom-4'-methyloacetophenone ≥98.0%
3'-Brom-4'-methyloacetophenone
Formel:
C₉H₉BrO MW: 213,0700 g/mol Kokpunkt: 118°C |
MDL Number:
MFCD02683854 CAS nummer: 40180-80-1 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 42.0 to 46.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...