N-(4-Bromophenyl)-4-biphenylamine ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-(4-Bromoanilino)biphenyl
B4753-1G
B4753-5G
TCIAB4753-1GEA
885
SEK
TCIAB4753-1G
TCIAB4753-5G
N-(4-Bromophenyl)-4-biphenylamine ≥98.0%
N-(4-Bromophenyl)-4-biphenylamine
Formel:
C₁₈H₁₄BrN MW: 324,22 g/mol |
MDL Number:
MFCD20486764 CAS nummer: 1160294-93-8 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 130.0 to 134.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...