1,4-Bis(dimethylsilyl)benzene ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1,4-Phenylenebis(dimethylsilane)

B4748-5ML B4748-25ML
TCIAB4748-5MLEA 445 SEK
TCIAB4748-5ML TCIAB4748-25ML
1,4-Bis(dimethylsilyl)benzene ≥98.0%
1,4-Bis(dimethylsilyl)benzene
Formel: C₁₀H₁₈Si₂
MW: 194,42 g/mol
Kokpunkt: 220°C
Flampunkt: 85°C
MDL Number: MFCD00039790
CAS nummer: 2488-01-9
EINECS: 219-638-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f343304566eb05899db609d63eff07e51a43d83c&vid=f343304566eb05899db609d63eff07e51a43d83c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR