4'-Bromovalerophenone ≥98.0%

Leverantör: TCI EUROPE

B4733-5G B4733-25G
TCIAB4733-5GEA 285 SEK
TCIAB4733-5G TCIAB4733-25G
4'-Bromovalerophenone ≥98.0%
4'-Bromovalerophenone
Formel: C₁₁H₁₃BrO
MW: 241,13 g/mol
Kokpunkt: 170°C
Storage Temperature: Kylskåp
MDL Number: MFCD00000107
CAS nummer: 7295-44-5
EINECS: 230-729-1

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 36.0 to 40.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR