Basic Violet 8
Leverantör: TCI EUROPE
B4731-5G
B4731-25G
TCIAB4731-5GEA
270
SEK
TCIAB4731-5G
TCIAB4731-25G
Basic Violet 8
Basic Violet 8
CAS nummer:
8004-94-2 EINECS: 232-332-9 |
Specifikation testresultat
Appearance | Red to Dark blue to Black powder to crystal |
Lambda max. | 540.0 to 550.0 nm (H2O) |
Absorbance (E1%1cm) | min. 800 (H2O, 540.0 to 550.0 nm) |
Drying loss | max. 10.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...