2-Bromo-4-methoxy-6-nitrobenzenol ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
2-Bromo-4-methoxy-6-nitrophenol
B4723-1G
B4723-5G
TCIAB4723-1GEA
255
SEK
TCIAB4723-1G
TCIAB4723-5G
2-Bromo-4-methoxy-6-nitrobenzenol ≥98.0%
2-Bromo-4-methoxy-6-nitrobenzenol
Formel:
C₇H₆BrNO₄ MW: 248,04 g/mol |
MDL Number:
MFCD06797984 CAS nummer: 115929-59-4 |
Specifikation testresultat
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 116.0 to 120.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...