1-Bromo-2,4,5-trimethoxybenzene ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
B4721-200MG
B4721-1G
TCIAB4721-200MGEA
645
SEK
TCIAB4721-200MG
TCIAB4721-1G
1-Bromo-2,4,5-trimethoxybenzene ≥98.0%
1-Bromo-2,4,5-trimethoxybenzene
Formel:
C₉H₁₁BrO₃ MW: 247,09 g/mol Kokpunkt: 263°C |
MDL Number:
MFCD00094684 CAS nummer: 20129-11-7 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 53.0 to 57.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...