4-Bromo-3-nitrobenzonitrile ≥98.0%

Leverantör: TCI
Varning

B4708-1G B4708-5G
TCIAB4708-1GEA 390 SEK
TCIAB4708-1G TCIAB4708-5G
4-Bromo-3-nitrobenzonitrile ≥98.0%
4-Bromo-3-nitrobenzonitrile
Formel: C₇H₃BrN₂O₂
Storage Temperature: Kylskåp
MDL Number: MFCD00982006
CAS nummer: 89642-49-9

Order Now

Specifikation testresultat

Purity (GC) min. 98.0 %
Melting point 119.0 to 123.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR