3'-Benzyloxyacetophenone ≥98.0%
Leverantör: TCI EUROPE
Synonyms: 1-(3-(Benzyloxy)phenyl)ethanone, 1-[3-(Benzyloxy)phenyl]ethan-1-one
B4705-5G B4705-25G
TCIAB4705-5GEA 550 SEK
TCIAB4705-5G TCIAB4705-25G
3'-Benzyloxyacetophenone ≥98.0%
3'-Benzyloxyacetophenone
Formel: C₁₅H₁₄O₂
MW: 226,27 g/mol
Kokpunkt: 154°C
MDL Number: MFCD00026221
CAS nummer: 34068-01-4
EINECS: 251-816-0
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %