6-Bromo-4-hydroxycoumarin 98,0 GC, T

Leverantör: TCI
Varning

B4699-1G B4699-5G
TCIAB4699-1GEA 855 SEK
TCIAB4699-1G TCIAB4699-5G
6-Bromo-4-hydroxycoumarin 98,0 GC, T
6-Bromo-4-hydroxycoumarin
Formel: C₉H₅BrO₃
MW: 241.04 g/mol
Smältpunkt: 277°C
MDL Number: MFCD00239372
CAS nummer: 4139-61-1

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR