3-Bromo-2-nitropyridine ≥98.0%

Leverantör: TCI
Varning

B4690-1G B4690-5G
TCIAB4690-1GEA 495 SEK
TCIAB4690-1G TCIAB4690-5G
3-Bromo-2-nitropyridine ≥98.0%
3-Bromo-2-nitropyridine
Formel: C₅H₃BrN₂O₂
MW: 203 g/mol
MDL Number: MFCD00955614
CAS nummer: 54231-33-3

Order Now

Specifikation testresultat

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 102.0 to 106.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR