Bis(NHS)PEG₉

Leverantör: TCI

Synonyms: Bis(NHS)PEG9, 4,7,10,13,16,19,22,25,28-Nonaoxahentriacontanedioic acid di(N-succinimidyl) ester, Di(N-succinimidyl) 4,7,10,13,16,19,22,25,28-nonaoxahentriacontanedioate, 4,7,10,13,16,19,22,25,28-Nonaoxahentriacontanedioicacid di(N-succinimidyl) ester, Bis(2,5-dioxopyrrolidin-1-yl) 4,7,10,13,16,19,22,25,28-nonaoxahentriacontanedioate, Di(N-succinimidyl) 4,7,10,13,16,19,22,25,28-Nonaoxahentriacontanedioate, Bis-dPEG®9-NHS ester

B4688-25MG B4688-100MG
TCIAB4688-25MGEA 1325 SEK
TCIAB4688-25MG TCIAB4688-100MG
Bis(NHS)PEG₉
Bis(NHS)PEG9
Formel: C₃₀H₄₈N₂O₁₇
MW: 708.71 g/mol
Storage Temperature: Frys
MDL Number: MFCD11041131
CAS nummer: 1008402-79-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a90e9caab5c94c18988372f602d37bbc9a779728&vid=2d063f7bbbea433b5171186c9244f23b8abb348d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Elemental analysis (Nitrogen) 3.70 to 4.20 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR