4-Bromobenzhydrol ≥97.0%
Leverantör: TCI EUROPE
Synonyms:
p-Bromobenzhydrol, 4-Bromophenyl phenyl carbinol, 4-Bromodiphenylmethanol, 4-Bromo-α-phenylbenzyl alcohol, 4-Bromobenzhydryl alcohol, (4-Bromophenyl)(phenyl)methanol
B4680-1G
B4680-5G
TCIAB4680-1GEA
470
SEK
TCIAB4680-1G
TCIAB4680-5G
4-Bromobenzhydrol ≥97.0%
4-Bromobenzhydrol
Formel:
C₁₃H₁₁BrO MW: 263,13 g/mol |
MDL Number:
MFCD00016851 CAS nummer: 29334-16-5 EINECS: 249-568-3 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 63.0 to 67.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...