2'-Hydroxy-2-phenylacetophenone ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Benzyl 2-Hydroxyphenyl ketone, 2'-Hydroxy-2-phenylacetophenone, 1-(2-Hydroxyphenyl)-2-phenylethanone
B4673-200MG B4673-1G
TCIAB4673-200MGEA 845 SEK
TCIAB4673-200MG TCIAB4673-1G
2'-Hydroxy-2-phenylacetophenone ≥98.0%
2'-Hydroxy-2-phenylacetophenone
Formel: C₁₄H₁₂O₂
MW: 212,24 g/mol
Kokpunkt: 165°C
MDL Number: MFCD00218589
CAS nummer: 2491-31-8
Order Now
Specifikation testresultat
Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 55.0 to 59.0 deg-C