4-Bromo-9,9-dimethyl-9H-fluorene ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-Bromo-9,9-dimethylfluorene
B4661-200MG
B4661-1G
TCIAB4661-200MGEA
310
SEK
TCIAB4661-200MG
TCIAB4661-1G
4-Bromo-9,9-dimethyl-9H-fluorene ≥98.0%
4-Bromo-9,9-dimethyl-9H-fluorene
Formel:
C₁₅H₁₃Br |
MDL Number:
MFCD23099415 CAS nummer: 942615-32-9 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...