2-Bromo-3-pentanone ≥95.0%

Leverantör: TCI EUROPE
Varning

B4648-5G B4648-25G
TCIAB4648-5GEA 1475 SEK
TCIAB4648-5G TCIAB4648-25G
2-Bromo-3-pentanone ≥95.0%
2-Bromo-3-pentanone
Formel: C₅H₉BrO
MW: 165,03 g/mol
Kokpunkt: 55°C
Storage Temperature: Kylskåp
MDL Number: MFCD11036312
CAS nummer: 815-52-1
UN: 1224
ADR: 3,III

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR