3,3'-Bicarbazole ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI EUROPE
Varning

Synonyms: 9H,9'H-3,3'-Bicarbazole

B4646-200MG B4646-1G
TCIAB4646-200MGEA 870 SEK
TCIAB4646-200MG TCIAB4646-1G
3,3'-Bicarbazole ≥98.0% (by HPLC, total nitrogen)
3,3'-Bicarbazole
Formel: C₂₄H₁₆N₂
MW: 332,41 g/mol
MDL Number: MFCD11046470
CAS nummer: 1984-49-2

Order Now

Specifikation testresultat

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR