Benzo[b]thiophene-2-carbonyl chloride 98,0 Titrimetrisk
Leverantör: TCI
|
Fara
|
Synonyms:
Benzo[b]thiophene-2-carbonyl chloride
B4637-1G
B4637-5G
TCIAB4637-1GEA
670
SEK
TCIAB4637-1G
TCIAB4637-5G
Benzo[b]thiophene-2-carbonyl chloride 98,0 Titrimetrisk
Benzo[b]thiophene-2-carbonyl chloride
Formel:
C₉H₅ClOS MW: 196.65 g/mol Kokpunkt: 176°C |
MDL Number:
MFCD00051635 CAS nummer: 39827-11-7 UN: 3261 ADR: 8 II |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 83.0 to 88.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...