11H-Benzo[a]carbazole ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1,2-Benzocarbazole
B4635-1G
B4635-5G
TCIAB4635-1GEA
1425
SEK
TCIAB4635-1G
TCIAB4635-5G
11H-Benzo[a]carbazole ≥98.0%
11H-Benzo[a]carbazole
Formel:
C₁₆H₁₁N MW: 217,27 g/mol |
MDL Number:
MFCD00215939 CAS nummer: 239-01-0 EINECS: 205-945-4 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 231.0 to 235.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...