9,9-Bis(4-allyloxyphenyl)fluorene ≥97.0%

Leverantör: TCI EUROPE

Synonyms: 9,9-Bis(4-(allyloxy)phenyl)-9H-fluorene

B4622-5G B4622-25G
TCIAB4622-5GEA 1200 SEK
TCIAB4622-5G TCIAB4622-25G
9,9-Bis(4-allyloxyphenyl)fluorene ≥97.0%
9,9-Bis(4-allyloxyphenyl)fluorene
Formel: C₃₁H₂₆O₂
MW: 430,55 g/mol
MDL Number: MFCD28975099
CAS nummer: 142494-81-3

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 85.0 to 89.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR