4-(3-Bromophenyl)pyridine ≥98.0%

Leverantör: TCI EUROPE
Varning

B4616-1G B4616-5G
TCIAB4616-1GEA 1250 SEK
TCIAB4616-1G TCIAB4616-5G
4-(3-Bromophenyl)pyridine ≥98.0%
4-(3-Bromophenyl)pyridine
Formel: C₁₁H₈BrN
MW: 234,10 g/mol
MDL Number: MFCD16250222
CAS nummer: 4373-72-2

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 43.0 to 47.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR