2-bromo-5-methylthiazole ≥98.0%

Leverantör: TCI
Varning

Synonyms: 2-Bromo-5-methylthiazole

B4604-1G B4604-5G
TCIAB4604-1GEA 430 SEK
TCIAB4604-1G TCIAB4604-5G
2-bromo-5-methylthiazole ≥98.0%
2-bromo-5-methylthiazole
Formel: C₄H₄BrNS
MW: 178.047 g/mol
Kokpunkt: 84°C
MDL Number: MFCD08460610
CAS nummer: 41731-23-1

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR