4-Butylmorpholine ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Varning
B4602-5ML B4602-25ML
TCIAB4602-5MLEA 855 SEK
TCIAB4602-5ML TCIAB4602-25ML
4-Butylmorpholine ≥98.0% (by GC, titration analysis)
4-Butylmorpholine
Formel: C₈H₁₇NO
MW: 143.23 g/mol
Kokpunkt: 78°C
MDL Number: MFCD00023401
CAS nummer: 1005-67-0
UN: 1993
ADR: 3 III
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %