3-Bromo-5-fluorobenzonitrile ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
B4600-1G
B4600-5G
TCIAB4600-1GEA
295
SEK
TCIAB4600-1G
TCIAB4600-5G
3-Bromo-5-fluorobenzonitrile ≥98.0%
3-Bromo-5-fluorobenzonitrile
Formel:
C₇H₃BrFN MW: 200,01 g/mol |
MDL Number:
MFCD04038227 CAS nummer: 179898-34-1 UN: 3439 ADR: 6.1,III |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 44.0 to 48.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...