2-(Benzyloxy)phenylboronic acid (contains varying amounts of Anhydride)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
2-Benzyloxyphenylboronic acid, (2-(Benzyloxy)phenyl)boronic acid, 2-Benzyloxybenzeneboronic acid
B4590-1G
B4590-5G
TCIAB4590-1GEA
375
SEK
TCIAB4590-1G
TCIAB4590-5G
2-(Benzyloxy)phenylboronic acid (contains varying amounts of Anhydride)
2-(Benzyloxy)phenylboronic acid
Formel:
C₁₃H₁₃BO₃ MW: 228,06 g/mol Smältpunkt: 110°C |
MDL Number:
MFCD01632206 CAS nummer: 190661-29-1 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 109.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...