2-Bromo-3-(2-ethylhexyl)-5-iodothiophene ≥97.0% stabilised
Leverantör: TCI EUROPE
Stabilization: Stabilised with copper chip
B4585-1G B4585-5G
TCIAB4585-1GEA 910 SEK
TCIAB4585-1G TCIAB4585-5G
2-Bromo-3-(2-ethylhexyl)-5-iodothiophene ≥97.0% stabilised
2-Bromo-3-(2-ethylhexyl)-5-iodothiophene
Formel: C₁₂H₁₈BrIS
MW: 401,14 g/mol
Storage Temperature: Frys
CAS nummer: 1034352-30-1
Order Now
Specifikation testresultat
Purity (GC) min. 97.0 %