1-(Bromomethyl)naphthalene ≥98.0%

Leverantör: TCI EUROPE
Fara

B4563-5G B4563-25G
TCIAB4563-5GEA 550 SEK
TCIAB4563-5G TCIAB4563-25G
1-(Bromomethyl)naphthalene ≥98.0%
1-(Bromomethyl)naphthalene
Formel: C₁₁H₉Br
MW: 221,10 g/mol
Kokpunkt: 118°C
Storage Temperature: Kylskåp
MDL Number: MFCD00010804
CAS nummer: 3163-27-7
UN: 3261
ADR: 8,II

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 52.0 to 56.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR