2H-1,4-Benzothiazin-3(4H)-one ≥98.0%
Leverantör: TCI EUROPE
Synonyms: 2H-Benzo[b][1,4]thiazin-3(4H)-one
B4554-1G B4554-5G B4554-25G
TCIAB4554-1GEA 375 SEK
TCIAB4554-1G TCIAB4554-5G TCIAB4554-25G
2H-1,4-Benzothiazin-3(4H)-one ≥98.0%
2H-1,4-Benzothiazin-3(4H)-one
Formel: C₈H₇NOS
MW: 165,21 g/mol
MDL Number: MFCD00006698
CAS nummer: 5325-20-2
EINECS: 226-197-5
Order Now
Specifikation testresultat
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 178.0 to 182.0 deg-C