N,N-Bis(4-biphenylyl)-N-(4-bromophenyl)amine ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI EUROPE
Synonyms:
4-Bromo-4',4''-diphenyltriphenylamine
B4550-5G
B4550-25G
TCIAB4550-5GEA
975
SEK
TCIAB4550-5G
TCIAB4550-25G
N,N-Bis(4-biphenylyl)-N-(4-bromophenyl)amine ≥98.0% (by HPLC, total nitrogen)
N,N-Bis(4-biphenylyl)-N-(4-bromophenyl)amine
Formel:
C₃₀H₂₂BrN MW: 476,42 g/mol |
MDL Number:
MFCD20261458 CAS nummer: 499128-71-1 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 169.0 to 173.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...