7-(Bromomethyl)pentadecane ≥95.0%

Leverantör: TCI EUROPE
Varning

B4544-5ML B4544-25ML
TCIAB4544-5MLEA 965 SEK
TCIAB4544-5ML TCIAB4544-25ML
7-(Bromomethyl)pentadecane ≥95.0%
7-(Bromomethyl)pentadecane
Formel: C₁₆H₃₃Br
MW: 305,34 g/mol
Kokpunkt: 118°C
Flampunkt: 108°C
MDL Number: MFCD00136231
CAS nummer: 52997-43-0

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR