2,5-Bis(trimethylstannyl)thieno[3,2-b]thiophene ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
B4536-200MG
B4536-1G
TCIAB4536-200MGEA
1100
SEK
TCIAB4536-200MG
TCIAB4536-1G
2,5-Bis(trimethylstannyl)thieno[3,2-b]thiophene ≥98.0%
2,5-Bis(trimethylstannyl)thieno[3,2-b]thiophene
Formel:
C₁₂H₂₀S₂Sn₂ MW: 465,83 g/mol |
MDL Number:
MFCD22200503 CAS nummer: 469912-82-1 UN: 3146 ADR: 6.1,I |
Specifikation testresultat
Appearance | White to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 127.0 to 131.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...