1-Boc-3-cyanopiperidine ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: tert-butyl 3-cyanopiperidine-1-carboxylate, 1-Boc-3-cyanopiperidine, 3-Cyano-1-piperidinecarboxylic acid tert-Butyl ester, tert-Butyl 3-Cyano-1-piperidinecarboxylate, 1-Boc-3-piperidinecarbonitrile, 1-tert-Butoxycarbonyl-3-piperidinecarbonitrile, 1-tert-Butoxycarbonyl-3-cyanopiperidine

B4515-1G B4515-5G
TCIAB4515-1GEA 525 SEK
TCIAB4515-1G TCIAB4515-5G
1-Boc-3-cyanopiperidine ≥98.0%
1-Boc-3-cyanopiperidine
Formel: C₁₁H₁₈N₂O₂
MW: 210,27 g/mol
Kokpunkt: 40°C
Storage Temperature: Kylskåp
MDL Number: MFCD01861222
CAS nummer: 91419-53-3
UN: 3439
ADR: 6.1,III

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 65.0 to 69.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR