1-Bromo-4-fluoro-2-iodobenzene ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
B4506-5G
B4506-25G
TCIAB4506-5GEA
575
SEK
TCIAB4506-5G
TCIAB4506-25G
1-Bromo-4-fluoro-2-iodobenzene ≥98.0%
1-Bromo-4-fluoro-2-iodobenzene
Formel:
C₆H₃BrFI MW: 300,9 g/mol |
MDL Number:
MFCD00042578 CAS nummer: 202865-72-3 |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...