(2-Bromoethoxy)-tert-butyldimethylsilane ≥96.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
(2-Bromoethoxy)(tert-butyl)dimethylsilane
B4505-5G
B4505-25G
TCIAB4505-5GEA
310
SEK
TCIAB4505-5G
TCIAB4505-25G
(2-Bromoethoxy)-tert-butyldimethylsilane ≥96.0%
(2-Bromoethoxy)-tert-butyldimethylsilane
Formel:
C₈H₁₉BrOSi MW: 239,228 g/mol Kokpunkt: 120°C Flampunkt: 73°C |
MDL Number:
MFCD00209550 CAS nummer: 86864-60-0 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...