Boc-Tic-OH ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: (S)-N-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, Boc-Tic-OH, (S)-2-(tert-Butoxycarbonyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, Boc-L-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, (S)-2-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

B4501-5G B4501-25G
TCIAB4501-5GEA 725 SEK
TCIAB4501-5G TCIAB4501-25G
Boc-Tic-OH ≥98.0% (by HPLC, titration analysis)
Boc-Tic-OH
Formel: C₁₅H₁₉NO₄
MW: 277,32 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00143845
CAS nummer: 78879-20-6

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 129.0 to 133.0 deg-C
Specific rotation [a]20/D +19.0 to +22.0 deg (C=5,MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR