1-(4-Bromophenyl)-2-thiourea 98,0 HPLC, N
Leverantör: TCI
Fara
Synonyms: 1-(4-Bromophenyl)thiourea, N-(4-Bromophenyl)thiourea, 4-Bromophenylthiourea
B4484-1G B4484-5G
TCIAB4484-1GEA 710 SEK
TCIAB4484-1G TCIAB4484-5G
1-(4-Bromophenyl)-2-thiourea 98,0 HPLC, N
1-(4-Bromophenyl)-2-thiourea
Formel: (C₆H₄Br)NHC(S)NH₂
MW: 231.12 g/mol
Kokpunkt: 319
Smältpunkt: 186
MDL Number: MFCD00041176
CAS nummer: 2646-30-2
UN: 2811
ADR: 6.1 III
Order Now
Specifikation testresultat
Appearance White to Light yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 174.0 to 178.0 deg-C