Desyl bromide ≥95.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 2-Bromo-2-phenylacetophenone

B4471-5G B4471-25G
TCIAB4471-5GEA 1050 SEK
TCIAB4471-5G TCIAB4471-25G
Desyl bromide ≥95.0%
Desyl bromide
Formel: C₁₄H₁₁BrO
MW: 275,14 g/mol
MDL Number: MFCD00000136
CAS nummer: 1484-50-0
EINECS: 216-057-1
UN: 3261
ADR: 8,II

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 95.0 %
Melting point 54.0 to 58.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR