trans,trans-1,5-Bis(4-methoxyphenyl)-1,4-pentadien-3-one ≥98.0%

Leverantör: TCI

Synonyms: trans,trans-Bis(4-methoxybenzylidene)acetone, trans,trans-Bis(4-methoxybenzal)acetone, trans,trans-Bis(4-methoxystyryl) Ketone

B4467-200MG B4467-1G
TCIAB4467-200MGEA 485 SEK
TCIAB4467-200MG TCIAB4467-1G
trans,trans-1,5-Bis(4-methoxyphenyl)-1,4-pentadien-3-one ≥98.0%
trans,trans-1,5-Bis(4-methoxyphenyl)-1,4-pentadien-3-one
Formel: C₁₉H₁₈O₃
MDL Number: MFCD00014901
CAS nummer: 37951-12-5

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 127.0 to 131.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR