5-Bromo-2-methoxy-3-nitropyridine ≥96.0%
Leverantör: TCI EUROPE
|
Varning
|
B4461-1G
B4461-5G
TCIAB4461-1GEA
215
SEK
TCIAB4461-1G
TCIAB4461-5G
5-Bromo-2-methoxy-3-nitropyridine ≥96.0%
5-Bromo-2-methoxy-3-nitropyridine
Formel:
C₆H₅BrN₂O₃ MW: 233,0200 g/mol |
MDL Number:
MFCD07374968 CAS nummer: 152684-30-5 |
Specifikation testresultat
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 88.0 to 92.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...