2-Bromodibenzofuran ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 2-Bromodibenzo[b,d]furan
B4459-1G B4459-5G
TCIAB4459-1GEA 215 SEK
TCIAB4459-1G TCIAB4459-5G
2-Bromodibenzofuran ≥98.0%
2-Bromodibenzofuran
Formel: C₁₂H₇BrO
MW: 247,09 g/mol
Kokpunkt: 220°C
MDL Number: MFCD00092338
CAS nummer: 86-76-0
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 110.0 to 114.0 deg-C