6-Bromobenzimidazole ≥98.0%

Leverantör: TCI
Varning

Synonyms: 6-Bromo-1H-benzimidazole

B4458-1G B4458-5G
TCIAB4458-1GEA 455 SEK
TCIAB4458-1G TCIAB4458-5G
6-Bromobenzimidazole ≥98.0%
6-Bromobenzimidazole
Formel: C₇H₅BrN₂
MDL Number: MFCD00460014
CAS nummer: 4887-88-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9be849b0dbfc316cc9808b4f5331218afdc69834&vid=5726d2110984a83df916ee6a9d78640579bc02e2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 128.0 to 132.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR