2-Bromo-9,9-diphenylfluorene ≥98.0%

Leverantör: TCI

Synonyms: 2-Bromo-9,9-diphenyl-9H-fluorene

B4450-1G B4450-5G
TCIAB4450-1GEA 245 SEK
TCIAB4450-1G TCIAB4450-5G
2-Bromo-9,9-diphenylfluorene ≥98.0%
2-Bromo-9,9-diphenylfluorene
Formel: C₂₅H₁₇Br
MW: 397.32 g/mol
MDL Number: MFCD09907957
CAS nummer: 474918-32-6

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 217.0 to 221.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR